Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
Chromosome Location
Chr.9: 136940433 - 136944774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018998.3 NP_061871.1 5 - 6 856 57
XM_005266089.2 XP_005266146.2 5 - 6 906 57
XM_011518789.1 XP_011517091.1 5 - 6 906 57
XM_017014812.1 XP_016870301.1 5 - 6 876 57
GenBank mRNA AB209726.1 - 2 - 3 268 57
AK026081.1 - 5 - 6 720 57
AK315738.1 - 5 - 6 731 57
AL137631.1 - 1 - 2 119 57
BC004541.2 - 2 - 3 265 57
BC014130.1 - 5 - 6 752 57
BC014297.2 - 5 - 6 723 57
BC031100.1 - 4 - 5 871 57

View Full Product Details