The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008563.4 | NP_001008563.2 | 15 - 16 | 1973 | 66 |
NM_001110303.3 | NP_001103773.2 | 15 - 16 | 1875 | 66 | |
NM_006676.7 | NP_006667.3 | 15 - 16 | 1875 | 66 | |
XM_005251665.3 | XP_005251722.1 | 15 - 16 | 1863 | 66 | |
XM_005251666.3 | XP_005251723.1 | 10 - 11 | 1437 | 66 | |
XM_011518161.2 | XP_011516463.1 | 16 - 17 | 2215 | 66 | |
XM_011518162.2 | XP_011516464.1 | 15 - 16 | 2394 | 66 | |
XM_017014221.1 | XP_016869710.1 | 10 - 11 | 1437 | 66 | |
GenBank mRNA | AB023220.1 | - | 14 - 15 | 1793 | 66 |
AY074877.1 | - | 15 - 16 | 1909 | 66 | |
BC039593.1 | - | 15 - 16 | 1819 | 66 |