The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008563.4 | NP_001008563.2 | 4 - 5 | 442 | 63 |
NM_001110303.3 | NP_001103773.2 | 4 - 5 | 344 | 63 | |
NM_006676.7 | NP_006667.3 | 4 - 5 | 344 | 63 | |
XM_005251665.3 | XP_005251722.1 | 4 - 5 | 332 | 63 | |
XM_011518161.2 | XP_011516463.1 | 5 - 6 | 684 | 63 | |
XM_011518162.2 | XP_011516464.1 | 4 - 5 | 863 | 63 | |
GenBank mRNA | AB023220.1 | - | 3 - 4 | 265 | 63 |
AY074877.1 | - | 4 - 5 | 381 | 63 | |
BC039593.1 | - | 4 - 5 | 291 | 63 | |
BF727180.1 | - | 4 - 5 | 357 | 63 | |
BP246486.1 | - | 4 - 5 | 332 | 63 | |
DA791069.1 | - | 4 - 5 | 332 | 63 |