Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASA1
Entrez Gene ID
5921
Gene Name
RAS p21 protein activator 1
Gene Aliases
CM-AVM, CMAVM, GAP, PKWS, RASA, RASGAP, p120, p120GAP, p120RASGAP
Chromosome Location
Chr.5: 87267801 - 87391926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002890.2 NP_002881.1 1 - 2 740 78
XM_011543525.1 XP_011541827.1 1 - 2 1192 78
XM_011543527.2 XP_011541829.1 1 - 2 1192 78
GenBank mRNA AB209106.1 - 1 - 2 740 78
AK300269.1 - 1 - 2 67 78
BC033015.2 - 1 - 2 728 78
CR749722.1 - 1 - 2 1105 78
M23379.1 - 1 - 2 659 78

View Full Product Details