The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006729.4 | NP_006720.1 | 18 - 19 | 2548 | 149 |
NM_007309.3 | NP_009293.1 | 18 - 19 | 2548 | 149 | |
GenBank mRNA | AK002029.1 | - | 6 - 7 | 820 | 149 |
BC117414.1 | - | 19 - 20 | 2215 | 149 | |
BC143838.1 | - | 19 - 20 | 2215 | 149 | |
Y15908.1 | - | 18 - 19 | 2502 | 149 | |
Y15909.1 | - | 18 - 19 | 2502 | 149 |