The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320973.1 | NP_001307902.1 | 5 - 6 | 1315 | 76 |
NM_003666.3 | NP_003657.1 | 5 - 6 | 1315 | 76 | |
XM_011510092.2 | XP_011508394.1 | 5 - 6 | 1218 | 76 | |
XM_017002645.1 | XP_016858134.1 | 4 - 5 | 985 | 76 | |
GenBank mRNA | AK223533.1 | - | 5 - 6 | 1177 | 76 |
AK291725.1 | - | 5 - 6 | 982 | 76 | |
BC020716.1 | - | 5 - 6 | 964 | 76 | |
U79751.1 | - | 5 - 6 | 1204 | 76 |