Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNGR1
Entrez Gene ID
9145
Gene Name
synaptogyrin 1
Gene Aliases
-
Chromosome Location
Chr.22: 39349949 - 39385588 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004711.4 NP_004702.2 1 - 2 165 67
NM_145731.3 NP_663783.1 1 - 2 165 67
GenBank mRNA AJ002304.1 - 1 - 2 103 67
AJ002305.1 - 1 - 2 103 67
AK289507.1 - 1 - 2 112 67
AK295863.1 - 1 - 2 123 67
BC000731.2 - 1 - 2 129 67
BI667990.1 - 1 - 2 189 67
BT007135.1 - 1 - 2 103 67
CR456590.1 - 1 - 2 113 67

View Full Product Details