Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE3A
Entrez Gene ID
7337
Gene Name
ubiquitin protein ligase E3A
Gene Aliases
ANCR, AS, E6-AP, EPVE6AP, HPVE6A
Chromosome Location
Chr.15: 25337234 - 25439086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000462.3 NP_000453.2 8 - 9 2522 133
NM_130838.1 NP_570853.1 4 - 5 1737 133
NM_130839.2 NP_570854.1 7 - 8 2457 133
XM_005268267.4 XP_005268324.1 9 - 10 2824 133
XM_005268268.4 XP_005268325.1 8 - 9 2711 133
XM_005268269.4 XP_005268326.1 9 - 10 2555 133
XM_005268270.4 XP_005268327.1 8 - 9 2512 133
XM_005268271.4 XP_005268328.1 5 - 6 2331 133
XM_006720675.3 XP_006720738.1 9 - 10 2776 133
XM_006720676.3 XP_006720739.1 6 - 7 2021 133
XM_011521994.2 XP_011520296.1 9 - 10 2713 133
XM_011521995.2 XP_011520297.1 9 - 10 2387 133
XM_017022544.1 XP_016878033.1 10 - 11 2749 133
XM_017022545.1 XP_016878034.1 9 - 10 2708 133
XM_017022546.1 XP_016878035.1 9 - 10 2616 133
XM_017022547.1 XP_016878036.1 8 - 9 2648 133
XM_017022548.1 XP_016878037.1 8 - 9 2643 133
XM_017022549.1 XP_016878038.1 8 - 9 2450 133
XM_017022550.1 XP_016878039.1 8 - 9 2602 133
XM_017022551.1 XP_016878040.1 9 - 10 2653 133
XM_017022552.1 XP_016878041.1 9 - 10 2343 133
XM_017022553.1 XP_016878042.1 9 - 10 2658 133
XM_017022554.1 XP_016878043.1 5 - 6 2662 133
XM_017022555.1 XP_016878044.1 7 - 8 2431 133
XM_017022556.1 XP_016878045.1 8 - 9 2580 133
XM_017022557.1 XP_016878046.1 7 - 8 2515 133
XM_017022558.1 XP_016878047.1 8 - 9 2711 133
XM_017022559.1 XP_016878048.1 5 - 6 2331 133
GenBank mRNA AB194232.1 - 6 - 7 1833 133
AF273050.1 - 5 - 6 1792 133
AK130491.1 - 3 - 4 1466 133
AK291405.1 - 5 - 6 1883 133
AK292514.1 - 7 - 8 2067 133
BC002582.2 - 7 - 8 2410 133
BC009271.1 - 2 - 3 1014 133
L07557.1 - 4 - 5 1759 133
U84404.1 - 9 - 10 2279 133
X98031.1 - 5 - 6 1762 133
X98032.1 - 3 - 4 1693 133
X98033.1 - 5 - 6 1753 133

View Full Product Details