The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002087.3 | NP_002078.1 | 2 - 3 | 394 | 68 |
XM_005257253.1 | XP_005257310.1 | 2 - 3 | 439 | 68 | |
GenBank mRNA | AF055008.1 | - | 2 - 3 | 200 | 68 |
AK000607.1 | - | 2 - 3 | 178 | 68 | |
AK222522.1 | - | 2 - 3 | 178 | 68 | |
AY124489.1 | - | 1 - 2 | 156 | 68 | |
BC000324.2 | - | 2 - 3 | 357 | 68 | |
BC010577.2 | - | 1 - 2 | 148 | 68 | |
BT006844.1 | - | 1 - 2 | 138 | 68 | |
DC336362.1 | - | 2 - 3 | 394 | 68 | |
M75161.1 | - | 1 - 2 | 150 | 68 | |
X62320.1 | - | 2 - 3 | 178 | 68 |