The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002087.3 | NP_002078.1 | 4 - 5 | 606 | 82 |
XM_005257253.1 | XP_005257310.1 | 4 - 5 | 651 | 82 | |
GenBank mRNA | AF055008.1 | - | 4 - 5 | 412 | 82 |
AK023348.1 | - | 2 - 3 | 154 | 82 | |
AK222522.1 | - | 4 - 5 | 390 | 82 | |
AK296090.1 | - | 4 - 5 | 344 | 82 | |
AK303830.1 | - | 4 - 5 | 387 | 82 | |
AY124489.1 | - | 3 - 4 | 368 | 82 | |
BC000324.2 | - | 4 - 5 | 569 | 82 | |
BC010577.2 | - | 3 - 4 | 360 | 82 | |
BT006844.1 | - | 3 - 4 | 350 | 82 | |
M75161.1 | - | 3 - 4 | 362 | 82 | |
X62320.1 | - | 4 - 5 | 390 | 82 |