The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002087.3 | NP_002078.1 | 10 - 11 | 1435 | 121 |
XM_005257253.1 | XP_005257310.1 | 10 - 11 | 1480 | 121 | |
GenBank mRNA | AF055008.1 | - | 10 - 11 | 1241 | 121 |
AK000607.1 | - | 10 - 11 | 1219 | 121 | |
AK023348.1 | - | 7 - 8 | 723 | 121 | |
AK222522.1 | - | 10 - 11 | 1219 | 121 | |
AK296090.1 | - | 10 - 11 | 1173 | 121 | |
AK303830.1 | - | 7 - 8 | 745 | 121 | |
AY124489.1 | - | 9 - 10 | 1197 | 121 | |
BC010577.2 | - | 9 - 10 | 1189 | 121 | |
M75161.1 | - | 9 - 10 | 1191 | 121 | |
X62320.1 | - | 10 - 11 | 1219 | 121 |