The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271875.1 | NP_001258804.1 | 8 - 9 | 1446 | 104 |
NM_001292025.1 | NP_001278954.1 | 8 - 9 | 1751 | 104 | |
XM_006721900.2 | XP_006721963.1 | 8 - 9 | 2061 | 104 | |
XM_011524812.2 | XP_011523114.1 | 8 - 9 | 834 | 104 | |
XM_017024640.1 | XP_016880129.1 | 7 - 8 | 1332 | 104 | |
XM_017024641.1 | XP_016880130.1 | 9 - 10 | 1463 | 104 | |
GenBank mRNA | AK000746.1 | - | 8 - 9 | 1751 | 104 |
AK000977.1 | - | 7 - 8 | 1101 | 104 | |
BC056882.1 | - | 8 - 9 | 1446 | 104 | |
CR627470.1 | - | 3 - 4 | 244 | 104 |