The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144961.1 | NP_001138433.1 | 1 - 2 | 175 | 93 |
NM_005007.3 | NP_004998.3 | 1 - 2 | 175 | 93 | |
GenBank mRNA | AK291893.1 | - | 1 - 2 | 175 | 93 |
AK313888.1 | - | 1 - 2 | 110 | 93 | |
BC105064.1 | - | 1 - 2 | 113 | 93 | |
BC105068.1 | - | 1 - 2 | 113 | 93 | |
BC143671.1 | - | 1 - 2 | 113 | 93 | |
BM044744.1 | - | 1 - 2 | 157 | 93 | |
X77909.1 | - | 1 - 2 | 124 | 93 |