Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SI
Entrez Gene ID
6476
Gene Name
sucrase-isomaltase
Gene Aliases
-
Chromosome Location
Chr.3: 164978898 - 165078495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001041.3 NP_001032.2 42 - 43 4981 127
XM_011513078.2 XP_011511380.1 42 - 43 4985 127
GenBank mRNA BC115034.1 - 42 - 43 4994 127
BC116452.1 - 42 - 43 4994 127
BC116453.1 - 42 - 43 4994 127
BC132834.1 - 42 - 43 4974 127
BC132860.1 - 42 - 43 4974 127
X63597.1 - 42 - 43 4980 127

View Full Product Details