The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002533.3 | NP_002524.2 | 4 - 5 | 349 | 103 |
NM_206840.2 | NP_996671.1 | 3 - 4 | 275 | 103 | |
XM_011544196.1 | XP_011542498.1 | 4 - 5 | 332 | 103 | |
XM_011544198.1 | XP_011542500.1 | 3 - 4 | 287 | 103 | |
GenBank mRNA | AK297070.1 | - | 4 - 5 | 308 | 103 |
AK298826.1 | - | 3 - 4 | 227 | 103 | |
AW997091.1 | - | 1 - 2 | 94 | 103 | |
BC005897.1 | - | 4 - 5 | 308 | 103 | |
BP284587.1 | - | 4 - 5 | 349 | 103 | |
U68140.1 | - | 4 - 5 | 343 | 103 | |
U78772.1 | - | 3 - 4 | 256 | 103 |