The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201428.1 | NP_001188357.1 | 4 - 5 | 495 | 112 |
NM_015530.4 | NP_056345.3 | 4 - 5 | 1250 | 112 | |
XM_006712408.2 | XP_006712471.1 | 4 - 5 | 573 | 112 | |
GenBank mRNA | AK023082.1 | - | 4 - 5 | 522 | 112 |
AK023201.1 | - | 4 - 5 | 535 | 112 | |
AK027349.1 | - | 4 - 5 | 449 | 112 | |
AK125761.1 | - | 4 - 5 | 1250 | 112 | |
AK225810.1 | - | 4 - 5 | 532 | 112 | |
AK296698.1 | - | 5 - 6 | 559 | 112 | |
AK298020.1 | - | 4 - 5 | 529 | 112 | |
AK298771.1 | - | 2 - 3 | 116 | 112 | |
AL117430.1 | - | 4 - 5 | 461 | 112 | |
BC007770.2 | - | 4 - 5 | 484 | 112 |