The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201428.1 | NP_001188357.1 | 8 - 9 | 972 | 89 |
NM_015530.4 | NP_056345.3 | 8 - 9 | 1727 | 89 | |
XM_006712408.2 | XP_006712471.1 | 8 - 9 | 1050 | 89 | |
GenBank mRNA | AK023201.1 | - | 8 - 9 | 1012 | 89 |
AK027202.1 | - | 3 - 4 | 1179 | 89 | |
AK125761.1 | - | 8 - 9 | 1727 | 89 | |
AK225810.1 | - | 8 - 9 | 1009 | 89 | |
AK293640.1 | - | 7 - 8 | 859 | 89 | |
AK296698.1 | - | 9 - 10 | 1036 | 89 | |
AK298020.1 | - | 8 - 9 | 1006 | 89 | |
AK298771.1 | - | 6 - 7 | 593 | 89 | |
AK315508.1 | - | 8 - 9 | 1027 | 89 | |
AL117430.1 | - | 8 - 9 | 938 | 89 | |
BC007770.2 | - | 8 - 9 | 961 | 89 |