The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252613.1 | NP_001239542.1 | 1 - 2 | 120 | 98 |
NM_014497.4 | NP_055312.2 | 1 - 2 | 120 | 98 | |
XM_017003810.1 | XP_016859299.1 | 1 - 2 | 121 | 98 | |
GenBank mRNA | AK291198.1 | - | 1 - 2 | 84 | 98 |
BC083513.1 | - | 1 - 2 | 79 | 98 | |
BP348645.1 | - | 1 - 2 | 113 | 98 | |
D83032.1 | - | 1 - 2 | 112 | 98 |