The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320977.1 | NP_001307906.1 | 12 - 13 | 2287 | 122 |
NM_006122.3 | NP_006113.2 | 11 - 12 | 1978 | 122 | |
NR_135502.1 | 11 - 12 | 2051 | 122 | ||
NR_135503.1 | 11 - 12 | 2051 | 122 | ||
NR_135504.1 | 10 - 11 | 1776 | 122 | ||
XM_005254910.3 | XP_005254967.1 | 11 - 12 | 1926 | 122 | |
XM_011521565.2 | XP_011519867.1 | 5 - 6 | 818 | 122 | |
XM_011521566.2 | XP_011519868.1 | 5 - 6 | 818 | 122 | |
XM_017022184.1 | XP_016877673.1 | 11 - 12 | 1829 | 122 | |
XM_017022185.1 | XP_016877674.1 | 11 - 12 | 1926 | 122 | |
GenBank mRNA | AK289863.1 | - | 11 - 12 | 1873 | 122 |
AK293799.1 | - | 3 - 4 | 467 | 122 | |
AK295646.1 | - | 5 - 6 | 670 | 122 | |
AK308838.1 | - | 10 - 11 | 2120 | 122 | |
BC136448.1 | - | 10 - 11 | 1776 | 122 | |
BC144111.1 | - | 10 - 11 | 1776 | 122 | |
D55649.1 | - | 11 - 12 | 1826 | 122 | |
L28821.1 | - | 11 - 12 | 1826 | 122 |