The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320977.1 | NP_001307906.1 | 17 - 18 | 3005 | 132 |
NM_006122.3 | NP_006113.2 | 16 - 17 | 2696 | 132 | |
NR_135502.1 | 16 - 17 | 2794 | 132 | ||
NR_135503.1 | 16 - 17 | 2769 | 132 | ||
NR_135504.1 | 15 - 16 | 2519 | 132 | ||
XM_005254910.3 | XP_005254967.1 | 16 - 17 | 2644 | 132 | |
XM_011521565.2 | XP_011519867.1 | 10 - 11 | 1536 | 132 | |
XM_011521566.2 | XP_011519868.1 | 10 - 11 | 1536 | 132 | |
XM_017022184.1 | XP_016877673.1 | 16 - 17 | 2547 | 132 | |
XM_017022185.1 | XP_016877674.1 | 16 - 17 | 2644 | 132 | |
GenBank mRNA | AK289863.1 | - | 16 - 17 | 2591 | 132 |
AK293799.1 | - | 8 - 9 | 1185 | 132 | |
AK295646.1 | - | 10 - 11 | 1388 | 132 | |
AK316150.1 | - | 2 - 3 | 199 | 132 | |
AL832306.1 | - | 1 - 2 | 57 | 132 | |
BC136448.1 | - | 15 - 16 | 2494 | 132 | |
BC144111.1 | - | 15 - 16 | 2519 | 132 |