The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320977.1 | NP_001307906.1 | 4 - 5 | 914 | 73 |
NM_006122.3 | NP_006113.2 | 3 - 4 | 605 | 73 | |
NR_135502.1 | 3 - 4 | 678 | 73 | ||
NR_135503.1 | 3 - 4 | 678 | 73 | ||
NR_135504.1 | 2 - 3 | 403 | 73 | ||
XM_005254910.3 | XP_005254967.1 | 3 - 4 | 553 | 73 | |
XM_017022184.1 | XP_016877673.1 | 3 - 4 | 456 | 73 | |
XM_017022185.1 | XP_016877674.1 | 3 - 4 | 553 | 73 | |
GenBank mRNA | AK289863.1 | - | 3 - 4 | 500 | 73 |
AK308838.1 | - | 2 - 3 | 747 | 73 | |
BC136448.1 | - | 2 - 3 | 403 | 73 | |
BC144111.1 | - | 2 - 3 | 403 | 73 | |
D55649.1 | - | 3 - 4 | 453 | 73 | |
L28821.1 | - | 3 - 4 | 453 | 73 |