Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUN1
Entrez Gene ID
23353
Gene Name
Sad1 and UNC84 domain containing 1
Gene Aliases
UNC84A
Chromosome Location
Chr.7: 815557 - 874920 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX649069.1 - 3 - 4 435 66

View Full Product Details