The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143966.3 | NP_001137438.1 | 2 - 3 | 247 | 117 |
NM_001318806.1 | NP_001305735.1 | 2 - 3 | 247 | 117 | |
GenBank mRNA | AL833474.1 | - | 2 - 3 | 220 | 117 |
AY826820.1 | - | 2 - 3 | 173 | 117 | |
BC050465.1 | - | 2 - 3 | 189 | 117 | |
DA169008.1 | - | 2 - 3 | 342 | 117 |