The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320759.1 | NP_001307688.1 | 10 - 11 | 1518 | 73 |
NM_004618.4 | NP_004609.1 | 11 - 12 | 1592 | 73 | |
GenBank mRNA | AK296437.1 | - | 10 - 11 | 1350 | 73 |
BC051748.1 | - | 11 - 12 | 1474 | 73 | |
U43431.1 | - | 11 - 12 | 1455 | 73 |