The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003235.4 | NP_003226.4 | 1 - 2 | 109 | 83 |
XM_005251038.4 | XP_005251095.1 | 1 - 2 | 126 | 83 | |
XM_005251040.4 | XP_005251097.1 | 1 - 2 | 126 | 83 | |
XM_005251042.4 | XP_005251099.1 | 1 - 2 | 126 | 83 | |
XM_006716622.3 | XP_006716685.1 | 1 - 2 | 126 | 83 | |
XM_017013793.1 | XP_016869282.1 | 1 - 2 | 126 | 83 | |
XM_017013794.1 | XP_016869283.1 | 1 - 2 | 126 | 83 | |
XM_017013795.1 | XP_016869284.1 | 1 - 2 | 126 | 83 | |
XM_017013796.1 | XP_016869285.1 | 1 - 2 | 126 | 83 | |
XM_017013797.1 | XP_016869286.1 | 1 - 2 | 118 | 83 | |
XM_017013798.1 | XP_016869287.1 | 1 - 2 | 126 | 83 | |
XM_017013799.1 | XP_016869288.1 | 1 - 2 | 126 | 83 | |
XM_017013800.1 | XP_016869289.1 | 1 - 2 | 126 | 83 | |
GenBank mRNA | AK303666.1 | - | 1 - 2 | 111 | 83 |
BC140933.1 | - | 1 - 2 | 102 | 83 | |
U93033.2 | - | 1 - 2 | 68 | 83 | |
X02154.1 | - | 1 - 2 | 83 | ||
X05615.1 | - | 1 - 2 | 83 |