The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017829.5 | NP_060299.4 | 1 - 2 | 83 | 63 |
GenBank mRNA | AF273270.1 | - | 1 - 2 | 83 | 63 |