The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256324.1 | NP_001243253.1 | 13 - 14 | 3303 | 67 |
NM_001256325.1 | NP_001243254.1 | 13 - 14 | 3303 | 67 | |
NM_001256326.1 | NP_001243255.1 | 13 - 14 | 3303 | 67 | |
NM_001256327.1 | NP_001243256.1 | 13 - 14 | 3303 | 67 | |
NM_001256328.1 | NP_001243257.1 | 13 - 14 | 3303 | 67 | |
NM_001256329.1 | NP_001243258.1 | 13 - 14 | 3303 | 67 | |
NM_001256330.1 | NP_001243259.1 | 13 - 14 | 3303 | 67 | |
NM_001256331.1 | NP_001243260.1 | 13 - 14 | 3303 | 67 | |
NM_001256333.1 | NP_001243262.1 | 13 - 14 | 3303 | 67 | |
NM_001256334.1 | NP_001243263.1 | 13 - 14 | 3303 | 67 | |
NM_001256359.1 | NP_001243288.1 | 13 - 14 | 3303 | 67 | |
NM_001256360.1 | NP_001243289.1 | 13 - 14 | 3303 | 67 | |
NM_001256361.1 | NP_001243290.1 | 13 - 14 | 3303 | 67 | |
NM_018896.4 | NP_061496.2 | 13 - 14 | 3303 | 67 | |
NM_198377.2 | NP_938191.2 | 13 - 14 | 3303 | 67 | |
NM_198378.2 | NP_938192.1 | 13 - 14 | 3303 | 67 | |
NM_198380.2 | NP_938194.1 | 13 - 14 | 3303 | 67 | |
NM_198384.2 | NP_938198.1 | 13 - 14 | 3303 | 67 | |
NM_198385.2 | NP_938199.1 | 13 - 14 | 3303 | 67 | |
NM_198386.2 | NP_938200.1 | 13 - 14 | 3303 | 67 | |
NR_046054.1 | 13 - 14 | 3303 | 67 | ||
NR_046055.1 | 13 - 14 | 3303 | 67 | ||
NR_046056.1 | 13 - 14 | 3303 | 67 | ||
GenBank mRNA | AB012043.1 | - | 13 - 14 | 2939 | 67 |
AB032949.2 | - | 6 - 7 | 1265 | 67 | |
AF134986.1 | - | 13 - 14 | 2911 | 67 | |
AF227744.1 | - | 13 - 14 | 2911 | 67 | |
AF227745.1 | - | 13 - 14 | 2911 | 67 | |
AF227749.1 | - | 13 - 14 | 2911 | 67 | |
AF227750.1 | - | 13 - 14 | 2911 | 67 | |
AF227751.1 | - | 13 - 14 | 2911 | 67 | |
DQ494457.1 | - | 13 - 14 | 2911 | 67 | |
DQ494458.1 | - | 13 - 14 | 2911 | 67 | |
DQ494459.1 | - | 13 - 14 | 2911 | 67 | |
DQ494460.1 | - | 13 - 14 | 2911 | 67 | |
DQ494461.1 | - | 13 - 14 | 2911 | 67 | |
DQ494462.1 | - | 13 - 14 | 2911 | 67 | |
DQ494463.1 | - | 13 - 14 | 2911 | 67 | |
DQ494464.1 | - | 13 - 14 | 2911 | 67 | |
DQ494465.1 | - | 13 - 14 | 2911 | 67 | |
DQ494466.1 | - | 13 - 14 | 2911 | 67 | |
DQ494467.1 | - | 13 - 14 | 2911 | 67 | |
DQ494468.1 | - | 13 - 14 | 2911 | 67 | |
DQ494469.1 | - | 13 - 14 | 2911 | 67 | |
DQ494470.1 | - | 13 - 14 | 2911 | 67 | |
DQ494471.1 | - | 13 - 14 | 2911 | 67 | |
DQ494473.1 | - | 13 - 14 | 2911 | 67 | |
DQ494474.1 | - | 13 - 14 | 2911 | 67 | |
DQ494475.1 | - | 13 - 14 | 2911 | 67 | |
DQ494476.1 | - | 13 - 14 | 2911 | 67 | |
DQ494477.1 | - | 13 - 14 | 2911 | 67 | |
DQ494478.1 | - | 13 - 14 | 2911 | 67 | |
DQ494479.1 | - | 13 - 14 | 2911 | 67 |