The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081955.2 | NP_001075424.1 | 13 - 14 | 1169 | 63 |
NM_001165933.1 | NP_001159405.1 | 13 - 14 | 1169 | 63 | |
NM_003835.3 | NP_003826.2 | 13 - 14 | 1178 | 63 | |
XM_011525426.2 | XP_011523728.1 | 6 - 7 | 1349 | 63 | |
GenBank mRNA | AF071476.1 | - | 13 - 14 | 1042 | 63 |
AF073710.1 | - | 4 - 5 | 903 | 63 | |
AF493932.1 | - | 3 - 4 | 288 | 63 | |
AK289876.1 | - | 13 - 14 | 1042 | 63 | |
AK290535.1 | - | 13 - 14 | 1038 | 63 | |
AK301719.1 | - | 13 - 14 | 1048 | 63 | |
AK309061.1 | - | 13 - 14 | 1059 | 63 | |
AY585190.1 | - | 3 - 4 | 288 | 63 | |
AY585191.1 | - | 13 - 14 | 966 | 63 | |
BC022504.1 | - | 13 - 14 | 1036 | 63 | |
BC096843.1 | - | 3 - 4 | 291 | 63 |