Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS9
Entrez Gene ID
8787
Gene Name
regulator of G-protein signaling 9
Gene Aliases
PERRS, RGS9L
Chromosome Location
Chr.17: 65137338 - 65227703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF073710.1 - 1 - 2 615 103

View Full Product Details