The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002795.3 | NP_002786.2 | 5 - 6 | 675 | 74 |
NR_104194.1 | 4 - 5 | 497 | 74 | ||
NR_104195.1 | 5 - 6 | 598 | 74 | ||
GenBank mRNA | BC013008.2 | - | 5 - 6 | 638 | 74 |
BQ428285.1 | - | 5 - 6 | 558 | 74 | |
CN412852.1 | - | 4 - 5 | 401 | 74 | |
D26598.1 | - | 5 - 6 | 578 | 74 |