Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGRP3
Entrez Gene ID
25780
Gene Name
RAS guanyl releasing protein 3
Gene Aliases
GRP3
Chromosome Location
Chr.2: 33436348 - 33564731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139488.1 NP_001132960.1 17 - 18 2779 79
NM_015376.2 NP_056191.1 16 - 17 2334 79
NM_170672.2 NP_733772.1 18 - 19 2692 79
XM_005264246.3 XP_005264303.1 17 - 18 3116 79
XM_011532746.2 XP_011531048.1 18 - 19 2566 79
XM_011532747.2 XP_011531049.1 17 - 18 2581 79
XM_011532748.2 XP_011531050.2 17 - 18 2427 79
XM_017003759.1 XP_016859248.1 19 - 20 4042 79
XM_017003760.1 XP_016859249.1 18 - 19 4514 79
XM_017003761.1 XP_016859250.1 17 - 18 2444 79
XM_017003762.1 XP_016859251.1 18 - 19 2690 79
XM_017003763.1 XP_016859252.1 17 - 18 2420 79
GenBank mRNA AB020653.1 - 16 - 17 2334 79
AK292983.1 - 18 - 19 2603 79
BC027849.1 - 18 - 19 2692 79
BX647990.1 - 17 - 18 2387 79

View Full Product Details