Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSSC1
Entrez Gene ID
7260
Gene Name
tumor suppressing subtransferable candidate 1
Gene Aliases
-
Chromosome Location
Chr.2: 3188968 - 3377882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003310.2 NP_003301.1 6 - 7 847 85
XM_006711893.2 XP_006711956.1 4 - 5 752 85
XM_011510383.2 XP_011508685.1 7 - 8 983 85
XM_011510384.2 XP_011508686.1 7 - 8 876 85
XM_011510385.2 XP_011508687.1 6 - 7 876 85
XM_011510387.2 XP_011508689.1 5 - 6 638 85
XM_017004818.1 XP_016860307.1 6 - 7 948 85
GenBank mRNA AF019952.1 - 6 - 7 805 85
AK026522.1 - 6 - 7 788 85
AK222652.1 - 6 - 7 783 85
BC002485.2 - 6 - 7 755 85

View Full Product Details