Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSSC1
Entrez Gene ID
7260
Gene Name
tumor suppressing subtransferable candidate 1
Gene Aliases
-
Chromosome Location
Chr.2: 3188968 - 3377882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003310.2 NP_003301.1 7 - 8 1010 87
XM_006711893.2 XP_006711956.1 5 - 6 915 87
XM_011510383.2 XP_011508685.1 8 - 9 1146 87
XM_011510384.2 XP_011508686.1 8 - 9 1039 87
XM_011510385.2 XP_011508687.1 7 - 8 1039 87
XM_011510387.2 XP_011508689.1 6 - 7 801 87
XM_017004818.1 XP_016860307.1 7 - 8 1111 87
GenBank mRNA AF019952.1 - 7 - 8 968 87
AK026522.1 - 7 - 8 951 87
AK126288.1 - 10 - 11 1995 87
AK222652.1 - 7 - 8 946 87
BC002485.2 - 7 - 8 918 87

View Full Product Details