The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288653.1 | NP_001275582.1 | 28 - 29 | 4887 | 141 |
NM_004859.3 | NP_004850.1 | 28 - 29 | 4875 | 141 | |
XM_005257012.3 | XP_005257069.1 | 28 - 29 | 4687 | 141 | |
GenBank mRNA | AB073891.1 | - | 4 - 5 | 569 | 141 |
AK127134.1 | - | 28 - 29 | 4652 | 141 | |
BC051800.2 | - | 28 - 29 | 4644 | 141 | |
BC054489.1 | - | 28 - 29 | 4659 | 141 | |
BX640615.1 | - | 28 - 29 | 4659 | 141 | |
D21260.1 | - | 28 - 29 | 4604 | 141 |