Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MON1A
Entrez Gene ID
84315
Gene Name
MON1 homolog A, secretory trafficking associated
Gene Aliases
SAND1
Chromosome Location
Chr.3: 49908869 - 49930012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142501.1 NP_001135973.1 2 - 3 543 64
GenBank mRNA AK092018.1 - 2 - 3 342 64
BC047022.1 - 2 - 3 293 64

View Full Product Details