The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305102.1 | NP_001292031.1 | 10 - 11 | 1277 | 157 |
NM_007271.3 | NP_009202.1 | 10 - 11 | 1280 | 157 | |
XM_006714988.3 | XP_006715051.1 | 10 - 11 | 1660 | 157 | |
XM_006714989.3 | XP_006715052.1 | 10 - 11 | 1657 | 157 | |
XM_017010226.1 | XP_016865715.1 | 10 - 11 | 1240 | 157 | |
GenBank mRNA | AK312559.1 | - | 10 - 11 | 1188 | 157 |
BC012085.1 | - | 10 - 11 | 1234 | 157 | |
BC095413.1 | - | 10 - 11 | 1240 | 157 | |
Z35102.1 | - | 9 - 10 | 1543 | 157 |