The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162407.1 | NP_001155879.1 | 11 - 12 | 1677 | 84 |
NM_004071.3 | NP_004062.2 | 11 - 12 | 1397 | 84 | |
NR_027855.1 | 10 - 11 | 1306 | 84 | ||
NR_027856.1 | 9 - 10 | 2797 | 84 | ||
GenBank mRNA | AK025306.1 | - | 6 - 7 | 2704 | 84 |
AK294295.1 | - | 11 - 12 | 1677 | 84 | |
BC028149.1 | - | 10 - 11 | 1202 | 84 | |
BC031549.1 | - | 11 - 12 | 1293 | 84 | |
L29219.1 | - | 11 - 12 | 1371 | 84 | |
L29222.1 | - | 10 - 11 | 1280 | 84 | |
M59287.1 | - | 10 - 11 | 1281 | 84 |