The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032171.1 | NP_115547.1 | 14 - 15 | 2080 | 86 |
XM_005252265.2 | XP_005252322.1 | 14 - 15 | 2093 | 86 | |
XM_017015194.1 | XP_016870683.1 | 14 - 15 | 2090 | 86 | |
XM_017015195.1 | XP_016870684.1 | 14 - 15 | 2090 | 86 | |
GenBank mRNA | AK022705.1 | - | 14 - 15 | 2057 | 86 |
BC091515.1 | - | 2 - 3 | 443 | 86 |