Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
Chromosome Location
Chr.9: 114603897 - 114646423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153045.3 NP_694590.2 4 - 5 541 64
XM_006716997.1 XP_006717060.1 4 - 5 544 64
XM_011518350.1 XP_011516652.1 5 - 6 712 64
XM_011518351.2 XP_011516653.1 5 - 6 840 64
XM_011518352.1 XP_011516654.1 5 - 6 709 64
XM_011518353.1 XP_011516655.1 5 - 6 712 64
XM_011518354.1 XP_011516656.1 4 - 5 747 64
XM_011518355.1 XP_011516657.1 4 - 5 744 64
XM_017014429.1 XP_016869918.1 5 - 6 868 64
XM_017014430.1 XP_016869919.1 4 - 5 744 64
GenBank mRNA AK289383.1 - 4 - 5 510 64
AL557166.3 - 4 - 5 645 64
AL834412.1 - 2 - 3 153 64
BC107133.1 - 4 - 5 479 64
BC107134.1 - 4 - 5 692 64
BQ049157.1 - 4 - 5 610 64
BX649023.1 - 4 - 5 427 64

View Full Product Details