The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022158.3 | NP_071441.1 | 3 - 4 | 442 | 86 |
GenBank mRNA | AJ404615.1 | - | 3 - 4 | 402 | 86 |
AK054838.1 | - | 3 - 4 | 1392 | 86 | |
AK294871.1 | - | 3 - 4 | 403 | 86 | |
BC042680.1 | - | 3 - 4 | 386 | 86 |