Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FN3K
Entrez Gene ID
64122
Gene Name
fructosamine 3 kinase
Gene Aliases
-
Chromosome Location
Chr.17: 82735576 - 82751197 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022158.3 NP_071441.1 5 - 6 655 73
GenBank mRNA AJ404615.1 - 5 - 6 615 73
BC042680.1 - 5 - 6 599 73

View Full Product Details