Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBCD
Entrez Gene ID
6904
Gene Name
tubulin folding cofactor D
Gene Aliases
SSD-1, tfcD
Chromosome Location
Chr.17: 82752048 - 82943186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005993.4 NP_005984.3 21 - 22 2117 62
XM_005256396.4 XP_005256453.1 20 - 21 2082 62
XM_005256399.4 XP_005256456.1 9 - 10 914 62
XM_005256404.4 XP_005256461.1 9 - 10 956 62
XM_006722290.2 XP_006722353.1 20 - 21 2014 62
XM_006722291.3 XP_006722354.1 9 - 10 1238 62
XM_006722292.2 XP_006722355.1 7 - 8 650 62
XM_011523589.1 XP_011521891.1 16 - 17 1657 62
XM_011523591.1 XP_011521893.1 16 - 17 1642 62
XM_011523593.1 XP_011521895.1 15 - 16 1249 62
XM_011523594.1 XP_011521896.1 8 - 9 719 62
XM_011523595.2 XP_011521897.1 8 - 9 797 62
XM_011523597.2 XP_011521899.1 6 - 7 1450 62
XM_011523598.1 XP_011521900.1 6 - 7 508 62
XM_011523599.2 XP_011521901.1 6 - 7 1226 62
XM_011523600.2 XP_011521902.1 7 - 8 670 62
XM_017024987.1 XP_016880476.1 21 - 22 1978 62
XM_017024988.1 XP_016880477.1 21 - 22 2095 62
XM_017024989.1 XP_016880478.1 5 - 6 364 62
XM_017024990.1 XP_016880479.1 9 - 10 736 62
GenBank mRNA AB023205.1 - 21 - 22 2096 62
AF052173.1 - 7 - 8 505 62
AJ006417.1 - 21 - 22 2029 62
AK057975.1 - 6 - 7 624 62
AK091959.1 - 20 - 21 2003 62
AK293471.1 - 21 - 22 2095 62
AL133562.1 - 17 - 18 1562 62
BC003094.1 - 21 - 22 2117 62
BC006364.1 - 21 - 22 2074 62
BC039654.1 - 7 - 8 470 62
BC150142.1 - 5 - 6 436 62

View Full Product Details