The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134268.4 | NP_599030.1 | 2 - 3 | 741 | 58 |
XM_005257005.2 | XP_005257062.1 | 2 - 3 | 388 | 58 | |
XM_017024116.1 | XP_016879605.1 | 3 - 4 | 606 | 58 | |
GenBank mRNA | AB057769.1 | - | 2 - 3 | 421 | 58 |
AJ315162.1 | - | 2 - 3 | 378 | 58 | |
AK098057.1 | - | 2 - 3 | 388 | 58 | |
BC029798.1 | - | 2 - 3 | 417 | 58 |