The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032134.2 | NP_115510.1 | 11 - 12 | 4308 | 69 |
NR_130649.1 | 11 - 12 | 1399 | 69 | ||
XM_005257728.3 | XP_005257785.1 | 11 - 12 | 5318 | 69 | |
XM_006722136.3 | XP_006722199.1 | 7 - 8 | 1421 | 69 | |
XM_011525344.2 | XP_011523646.1 | 9 - 10 | 3991 | 69 | |
XM_017025206.1 | XP_016880695.1 | 11 - 12 | 5318 | 69 | |
XM_017025207.1 | XP_016880696.1 | 10 - 11 | 5258 | 69 | |
XM_017025208.1 | XP_016880697.1 | 9 - 10 | 1389 | 69 | |
GenBank mRNA | AL136774.1 | - | 11 - 12 | 4308 | 69 |
BC131559.1 | - | 11 - 12 | 1284 | 69 |