The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032134.2 | NP_115510.1 | 13 - 14 | 4497 | 82 |
NR_130649.1 | 13 - 14 | 1588 | 82 | ||
XM_005257728.3 | XP_005257785.1 | 13 - 14 | 5507 | 82 | |
XM_006722136.3 | XP_006722199.1 | 9 - 10 | 1610 | 82 | |
XM_011525344.2 | XP_011523646.1 | 11 - 12 | 4180 | 82 | |
XM_017025206.1 | XP_016880695.1 | 13 - 14 | 5507 | 82 | |
XM_017025207.1 | XP_016880696.1 | 12 - 13 | 5447 | 82 | |
XM_017025208.1 | XP_016880697.1 | 11 - 12 | 1578 | 82 | |
GenBank mRNA | AL136774.1 | - | 13 - 14 | 4497 | 82 |
BC131559.1 | - | 13 - 14 | 1473 | 82 |