The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198993.3 | NP_945344.1 | 8 - 9 | 1231 | 68 |
GenBank mRNA | AJ608762.1 | - | 8 - 9 | 1164 | 68 |
AK131500.1 | - | 8 - 9 | 1231 | 68 | |
AM903073.1 | - | 8 - 9 | 950 | 68 | |
BC109231.2 | - | 8 - 9 | 910 | 68 |