The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198993.3 | NP_945344.1 | 3 - 4 | 776 | 55 |
XM_017024580.1 | XP_016880069.1 | 3 - 4 | 776 | 55 | |
XM_017024581.1 | XP_016880070.1 | 3 - 4 | 776 | 55 | |
XM_017024582.1 | XP_016880071.1 | 3 - 4 | 776 | 55 | |
XM_017024583.1 | XP_016880072.1 | 3 - 4 | 776 | 55 | |
GenBank mRNA | AJ608762.1 | - | 3 - 4 | 709 | 55 |
AK131500.1 | - | 3 - 4 | 776 | 55 | |
AM903073.1 | - | 3 - 4 | 495 | 55 | |
BC109231.2 | - | 3 - 4 | 455 | 55 |