The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000418.3 | NP_000409.1 | 7 - 8 | 929 | 84 |
NM_001257406.1 | NP_001244335.1 | 6 - 7 | 796 | 84 | |
NM_001257407.1 | NP_001244336.1 | 7 - 8 | 1040 | 84 | |
NM_001257997.1 | NP_001244926.1 | 6 - 7 | 791 | 84 | |
XM_011545825.1 | XP_011544127.1 | 6 - 7 | 834 | 84 | |
XM_011545826.2 | XP_011544128.1 | 7 - 8 | 885 | 84 | |
XM_011545827.2 | XP_011544129.1 | 7 - 8 | 2124 | 84 | |
XM_011545828.2 | XP_011544130.1 | 5 - 6 | 602 | 84 | |
XM_011545829.1 | XP_011544131.1 | 6 - 7 | 763 | 84 | |
XM_011545830.2 | XP_011544132.1 | 6 - 7 | 1973 | 84 | |
XM_011545831.1 | XP_011544133.1 | 5 - 6 | 682 | 84 | |
XM_011545832.1 | XP_011544134.1 | 5 - 6 | 615 | 84 | |
XM_011545833.2 | XP_011544135.1 | 4 - 5 | 533 | 84 | |
XM_011545834.2 | XP_011544136.1 | 4 - 5 | 452 | 84 | |
XM_017023211.1 | XP_016878700.1 | 6 - 7 | 834 | 84 | |
GenBank mRNA | AK223591.1 | - | 7 - 8 | 847 | 84 |
AK303255.1 | - | 7 - 8 | 1015 | 84 | |
AK314863.1 | - | 6 - 7 | 781 | 84 | |
BC136365.1 | - | 6 - 7 | 828 | 84 | |
BC151131.1 | - | 6 - 7 | 828 | 84 | |
X52425.1 | - | 7 - 8 | 841 | 84 |