The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304514.1 | NP_001291443.1 | 10 - 11 | 1606 | 87 |
NM_022087.3 | NP_071370.2 | 11 - 12 | 1939 | 87 | |
XM_006716082.2 | XP_006716145.1 | 11 - 12 | 1850 | 87 | |
XM_006716083.2 | XP_006716146.1 | 11 - 12 | 1818 | 87 | |
XM_006716084.2 | XP_006716147.1 | 11 - 12 | 1782 | 87 | |
XM_017012500.1 | XP_016867989.1 | 10 - 11 | 1796 | 87 | |
GenBank mRNA | AK026056.1 | - | 7 - 8 | 1012 | 87 |
AK124934.1 | - | 13 - 14 | 2127 | 87 | |
AK298911.1 | - | 10 - 11 | 1529 | 87 | |
Y12434.3 | - | 10 - 11 | 1690 | 87 |