Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAN2A2
Entrez Gene ID
4122
Gene Name
mannosidase alpha class 2A member 2
Gene Aliases
MANA2X
Chromosome Location
Chr.15: 90902218 - 90922585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320977.1 NP_001307906.1 13 - 14 2401 118
NM_006122.3 NP_006113.2 12 - 13 2092 118
NR_135502.1 12 - 13 2165 118
NR_135503.1 12 - 13 2165 118
NR_135504.1 11 - 12 1890 118
XM_005254910.3 XP_005254967.1 12 - 13 2040 118
XM_011521565.2 XP_011519867.1 6 - 7 932 118
XM_011521566.2 XP_011519868.1 6 - 7 932 118
XM_017022184.1 XP_016877673.1 12 - 13 1943 118
XM_017022185.1 XP_016877674.1 12 - 13 2040 118
GenBank mRNA AK289863.1 - 12 - 13 1987 118
AK293799.1 - 4 - 5 581 118
AK295646.1 - 6 - 7 784 118
AK308838.1 - 11 - 12 2234 118
AL110288.1 - 2 - 3 128 118
BC136448.1 - 11 - 12 1890 118
BC144111.1 - 11 - 12 1890 118
D55649.1 - 12 - 13 1940 118
L28821.1 - 12 - 13 1940 118

View Full Product Details